COM448673016 Combi-Blocks, Inc.

3-(3-Methylphenoxy)propan-1-amine, HCl

  • MDL number MFCD07690524
  • Cas Number : 1052077-26-5
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3493
Availability details for
Quantity
  • Availability
  • Add to Cart