COM448673017 Combi-Blocks, Inc.

2-(4-Methoxy-phenoxy)-propylamine, HCl

  • MDL number MFCD03002050
  • Cas Number : 1352305-22-6
  • Molecular Weight : 217.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3494

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3494
Availability details for
Quantity
  • Availability
  • Add to Cart