COM448673107 Combi-Blocks, Inc.

2-(4-Methoxyphenyl)-2-morpholinoacetic acid

  • MDL number MFCD09026149
  • Cas Number : 803637-78-7
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3585

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3585
Availability details for
Quantity
  • Availability
  • Add to Cart