COM448673279 Combi-Blocks, Inc.

N-(4-Piperidinyl)acetamide, HCl

  • MDL number MFCD11870094
  • Cas Number : 58083-34-4
  • Molecular Weight : 178.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3759

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3759
Availability details for
Quantity
  • Availability
  • Add to Cart