COM448673445 Combi-Blocks, Inc.

2-(6-Chloropyrazin-2-ylamino)acetic acid

  • MDL number MFCD12068353
  • Cas Number : 1159822-45-3
  • Molecular Weight : 187.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3956

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3956
Availability details for
Quantity
  • Availability
  • Add to Cart