COM448673452 Combi-Blocks, Inc.

2-(6-Methoxynaphthalen-1-yl)propane-1,3-diol

  • MDL number MFCD09788622
  • Cas Number : 139293-28-0
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3966

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-3966
Availability details for
Quantity
  • Availability
  • Add to Cart