COM448673478 Combi-Blocks, Inc.

2-(Benzyloxycarbonylamino)-5-oxotetrahydrofuran-2-carboxylic acid

  • MDL number MFCD00546276
  • Cas Number : 104754-51-0
  • Molecular Weight : 279.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-4000

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-4000
Availability details for
Quantity
  • Availability
  • Add to Cart