COM448673824 Combi-Blocks, Inc.

4-(2-Hydroxyethyl)-piperazin-1-carboxylic acid ethyl ester

  • MDL number MFCD00191218
  • Cas Number : 14000-66-9
  • Molecular Weight : 202.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-4396

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-4396
Availability details for
Quantity
  • Availability
  • Add to Cart