COM448673863 Combi-Blocks, Inc.

4-(Trifluoromethylthio)phenylacetic acid

  • MDL number MFCD00236351
  • Cas Number : 243977-23-3
  • Molecular Weight : 236.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-4442

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-4442
Availability details for
Quantity
  • Availability
  • Add to Cart