COM448673887 Combi-Blocks, Inc.

Allyl 1,1,2,2-tetrafluoroethyl ether

  • MDL number MFCD00043822
  • Cas Number : 1428-33-7
  • Molecular Weight : 158.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-4471

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-4471
Availability details for
Quantity
  • Availability
  • Add to Cart