COM448674194 Combi-Blocks, Inc.

Pentafluorobenzenesulfonamide

  • MDL number MFCD02090183
  • Cas Number : 778-36-9
  • Molecular Weight : 247.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-4826

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-4826
Availability details for
Quantity
  • Availability
  • Add to Cart