COM448674246 Combi-Blocks, Inc.

2-Methyl-5-(trifluoromethyl)benzoyl chloride

  • MDL number MFCD01631599
  • Cas Number : 261952-08-3
  • Molecular Weight : 222.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-4888

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-4888
Availability details for
Quantity
  • Availability
  • Add to Cart