COM448674762 Combi-Blocks, Inc.

Ethyl 5-propyl-1h-pyrazole-4-carboxylate

  • MDL number MFCD01570118
  • Cas Number : 123374-28-7
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-5451

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-5451
Availability details for
Quantity
  • Availability
  • Add to Cart