COM448675417 Combi-Blocks, Inc.

3-(N-Prop-2-ynylcarbamoyl)prop-2-enoic acid

  • MDL number MFCD00111718
  • Cas Number : 671188-20-8
  • Molecular Weight : 153.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-6153

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-6153
Availability details for
Quantity
  • Availability
  • Add to Cart