COM448675996 Combi-Blocks, Inc.

3-[2-(Trifluoromethyl)phenyl]-1h-pyrazole

  • MDL number MFCD01569437
  • Cas Number : 149739-52-6
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-6756

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-6756
Availability details for
Quantity
  • Availability
  • Add to Cart