COM448676048 Combi-Blocks, Inc.

Hexakis(2,2,3,3-tetrafluoropropoxy)phosphazene

  • MDL number MFCD00798140
  • Cas Number : 58943-98-9
  • Molecular Weight : 921.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-6811

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-6811
Availability details for
Quantity
  • Availability
  • Add to Cart