COM448676288 Combi-Blocks, Inc.

1-[6-Fluoro-2-(trifluoromethyl)quinolin-4-yl]homopiperazine

  • MDL number MFCD03094525
  • Cas Number : 541539-67-7
  • Molecular Weight : 313.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7075

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7075
Availability details for
Quantity
  • Availability
  • Add to Cart