COM448676385 Combi-Blocks, Inc.

3,5-Dichloro-2,6-dimethoxybenzoic acid

  • MDL number MFCD00173910
  • Cas Number : 73219-91-7
  • Molecular Weight : 251.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7179

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7179
Availability details for
Quantity
  • Availability
  • Add to Cart