COM448676782 Combi-Blocks, Inc.

2-(Methylthio)-4-((tetrahydrofuran-2-yl)methylamino)pyrimidine-5-carboxylic acid

  • MDL number MFCD11100916
  • Cas Number : 1065075-56-0
  • Molecular Weight : 269.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7673

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7673
Availability details for
Quantity
  • Availability
  • Add to Cart