COM448676796 Combi-Blocks, Inc.

2-(Methylthio)-4-(4-(trifluoromethyl)phenylamino)pyrimidine-5-carboxylic acid

  • MDL number MFCD11100919
  • Cas Number : 1065075-64-0
  • Molecular Weight : 329.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7689

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7689
Availability details for
Quantity
  • Availability
  • Add to Cart