COM448676798 Combi-Blocks, Inc.

2-(Methylthio)-4-(phenethylamino)pyrimidine-5-carboxylic acid

  • MDL number MFCD11100918
  • Cas Number : 1065075-61-7
  • Molecular Weight : 289.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7691

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7691
Availability details for
Quantity
  • Availability
  • Add to Cart