COM448676942 Combi-Blocks, Inc.

3-Chloro-2,4-difluorobenzotrifluoride

  • MDL number MFCD04972709
  • Cas Number : 120770-03-8
  • Molecular Weight : 216.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7854

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7854
Availability details for
Quantity
  • Availability
  • Add to Cart