COM448676995 Combi-Blocks, Inc.

2-Methyl-1H-imidazole-4-carbaldehyde

  • MDL number MFCD00173725
  • Cas Number : 35034-22-1
  • Molecular Weight : 110.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-7920
Availability details for
Quantity
  • Availability
  • Add to Cart