COM448677109 Combi-Blocks, Inc.

4-Iodo-3,5-bis(trifluoromethyl)-1h-pyrazole

  • MDL number MFCD11867872
  • Cas Number : 1239511-06-8
  • Molecular Weight : 330

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8043

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8043
Availability details for
Quantity
  • Availability
  • Add to Cart