COM448677135 Combi-Blocks, Inc.

2-(N-Cbz-n-allylamino)-acetaldehyde

  • MDL number MFCD13184756
  • Cas Number : 370880-75-4
  • Molecular Weight : 233.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8073

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8073
Availability details for
Quantity
  • Availability
  • Add to Cart