COM448677137 Combi-Blocks, Inc.

2-(N-Isopropylmethylsulfonamido)acetic acid

  • MDL number MFCD09788596
  • Cas Number : 905587-49-7
  • Molecular Weight : 195.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8075

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8075
Availability details for
Quantity
  • Availability
  • Add to Cart