COM448677151 Combi-Blocks, Inc.

2-(Piperidin-4-ylthio)benzo[d]thiazole

  • MDL number MFCD02942421
  • Cas Number : 380637-96-7
  • Molecular Weight : 250.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8092

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8092
Availability details for
Quantity
  • Availability
  • Add to Cart