COM448677160 Combi-Blocks, Inc.

2-(Pyridin-2-yloxy)benzylamine, HCl

  • MDL number MFCD04117729
  • Cas Number : 870061-28-2
  • Molecular Weight : 236.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8107

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8107
Availability details for
Quantity
  • Availability
  • Add to Cart