COM448677189 Combi-Blocks, Inc.

2-(tert-Butoxycarbonyl-methyl-amino)-5-chloro-benzoic acid

  • MDL number MFCD03426355
  • Cas Number : 886362-04-5
  • Molecular Weight : 285.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8142

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8142
Availability details for
Quantity
  • Availability
  • Add to Cart