COM448677192 Combi-Blocks, Inc.

2-(tert-Butoxycarbonyl-phenyl-amino)-benzoic acid

  • MDL number MFCD03426358
  • Cas Number : 669713-65-9
  • Molecular Weight : 313.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8145

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8145
Availability details for
Quantity
  • Availability
  • Add to Cart