COM448677212 Combi-Blocks, Inc.

2,2,6,6-Tetramethyl-4-cyanopiperidine

  • MDL number MFCD07369663
  • Cas Number : 67845-90-3
  • Molecular Weight : 166.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8170

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8170
Availability details for
Quantity
  • Availability
  • Add to Cart