COM448677226 Combi-Blocks, Inc.

2,2-Diphenylethanesulfonyl chloride

  • MDL number MFCD02089493
  • Cas Number : 71351-01-4
  • Molecular Weight : 280.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8197

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8197
Availability details for
Quantity
  • Availability
  • Add to Cart