COM448677237 Combi-Blocks, Inc.

2,3,4,5-Tetrahydro-1h-benzo[e][1,4]diazepin-7-ylamine

  • MDL number MFCD07369990
  • Cas Number : 886366-79-6
  • Molecular Weight : 163.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8211

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8211
Availability details for
Quantity
  • Availability
  • Add to Cart