COM448677240 Combi-Blocks, Inc.

S-Benzyl-l-cysteinol

  • MDL number MFCD00235921
  • Cas Number : 85803-43-6
  • Molecular Weight : 197.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8215

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8215
Availability details for
Quantity
  • Availability
  • Add to Cart