COM448677257 Combi-Blocks, Inc.

2,3-Dihydro-1h-indene-1-methanamine

  • MDL number MFCD08457607
  • Cas Number : 54949-92-7
  • Molecular Weight : 147.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8249

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8249
Availability details for
Quantity
  • Availability
  • Add to Cart