COM448677272 Combi-Blocks, Inc.

2,3-Dimethoxy-1,4-benzoquinone

  • MDL number MFCD00016366
  • Cas Number : 3117-02-0
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8269

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8269
Availability details for
Quantity
  • Availability
  • Add to Cart