COM448677345 Combi-Blocks, Inc.

2,5-Dihydro-furan-2-carboxylic acid

  • MDL number MFCD07784085
  • Cas Number : 22694-55-9
  • Molecular Weight : 114.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8383

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8383
Availability details for
Quantity
  • Availability
  • Add to Cart