COM448677373 Combi-Blocks, Inc.

2,6-Dimethyl-d-tyrosine

  • MDL number MFCD07784105
  • Cas Number : 136771-16-9
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8431

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8431
Availability details for
Quantity
  • Availability
  • Add to Cart