COM448677387 Combi-Blocks, Inc.

2-[(1R)-1-Aminoethyl]-phenol

  • MDL number MFCD06761853
  • Cas Number : 123983-05-1
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8454

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8454
Availability details for
Quantity
  • Availability
  • Add to Cart