COM448677508 Combi-Blocks, Inc.

2-Amino-8-hydroxy-1,2,3,4-tetrahydro-naphthalene-2-carboxylic acid

  • MDL number MFCD07784024
  • Cas Number : 168628-97-5
  • Molecular Weight : 207.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8619

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8619
Availability details for
Quantity
  • Availability
  • Add to Cart