COM448677510 Combi-Blocks, Inc.

2-Amino-8-methoxy-1,2,3,4-tetrahydro-naphthalene-2-carboxylic acid

  • MDL number MFCD07369844
  • Cas Number : 67544-71-2
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8622

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8622
Availability details for
Quantity
  • Availability
  • Add to Cart