COM448677523 Combi-Blocks, Inc.

2-Aminomethyl-3-(4-bromo-phenyl)-propionic acid

  • MDL number MFCD07372594
  • Cas Number : 910443-87-7
  • Molecular Weight : 258.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8639

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8639
Availability details for
Quantity
  • Availability
  • Add to Cart