COM448677524 Combi-Blocks, Inc.

2-Aminomethyl-3-(4-methoxy-phenyl)-propionic acid

  • MDL number MFCD03002395
  • Cas Number : 682803-14-1
  • Molecular Weight : 209.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8641

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8641
Availability details for
Quantity
  • Availability
  • Add to Cart