COM448677550 Combi-Blocks, Inc.

2-Boc-6-iodo-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD12408121
  • Cas Number : 1008517-84-7
  • Molecular Weight : 359.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8681

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8681
Availability details for
Quantity
  • Availability
  • Add to Cart