COM448677557 Combi-Blocks, Inc.

2-Bromo-4,4'-di-tert-butyl-biphenyl

  • MDL number MFCD20921696
  • Cas Number : 70728-89-1
  • Molecular Weight : 345.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8695

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8695
Availability details for
Quantity
  • Availability
  • Add to Cart