COM448677561 Combi-Blocks, Inc.

2-Bromo-4-chloro-n-methoxy-n-methylbenzamide

  • MDL number MFCD17926350
  • Cas Number : 1255099-63-8
  • Molecular Weight : 278.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8701

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8701
Availability details for
Quantity
  • Availability
  • Add to Cart