COM448677566 Combi-Blocks, Inc.

2-Bromo-5-methoxy-3-methylbenzoic acid

  • MDL number MFCD09800403
  • Cas Number : 174417-54-0
  • Molecular Weight : 245.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8712

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8712
Availability details for
Quantity
  • Availability
  • Add to Cart