COM448677567 Combi-Blocks, Inc.

2-Bromo-5-methyl-3-nitro-thiophene

  • MDL number MFCD10687144
  • Cas Number : 82834-45-5
  • Molecular Weight : 222.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8713

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8713
Availability details for
Quantity
  • Availability
  • Add to Cart