COM448677603 Combi-Blocks, Inc.

2-Chloro-1-(3-chloro-phenyl)-ethanol

  • MDL number MFCD07784075
  • Cas Number : 106262-93-5
  • Molecular Weight : 191.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8771

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8771
Availability details for
Quantity
  • Availability
  • Add to Cart