COM448677608 Combi-Blocks, Inc.

2-Chloro-1-(4-chloro-phenyl)-ethanol

  • MDL number MFCD00462592
  • Cas Number : 6378-66-1
  • Molecular Weight : 191.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8777

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-8777
Availability details for
Quantity
  • Availability
  • Add to Cart